missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITSN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553702
This item is not returnable.
View return policy
Description
Immunogen sequence: ITVLEQQDMW WFGEVQGQKG WFPKSYVKLI SGPIRKSTSM DSGSSESPAS LKRVASPAAK PVVSGEEFIA MYTYESSEQG DLTFQQGDVI LVTKKDGDWW Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.
The protein encoded by this gene is a cytoplasmic membrane-associated protein that indirectly coordinates endocytic membrane traffic with the actin assembly machinery. In addition, the encoded protein may regulate the formation of clathrin-coated vesicles and could be involved in synaptic vesicle recycling. This protein has been shown to interact with dynamin, CDC42, SNAP23, SNAP25, SPIN90, EPS15, EPN1, EPN2, and STN2. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been characterized so far.
Specifications
| ITSN1 | |
| Polyclonal | |
| Unconjugated | |
| ITSN1 | |
| AA517634; AA545208; AI316805; AI839402; AI848451; EH and SH3 domains protein 1; EH domain and SH3 domain regulator of endocytosis 1; Eh domain, SH3 domain regulator of endocytosis 1; Ehsh1; Ese1; human intersectin-SH3 domain-containing protein SH3P17; intersectin (SH3 domain protein 1A); intersectin 1; intersectin 1 (SH3 domain protein 1A); intersectin 1 (SH3 domain protein); intersectin 1 short form variant 3; intersectin 1 short form variant, 11; intersectin 1; intersectin-1; intersectin short form 2 variant 2; intersectin short variant 12; intersectin-1; intersectin-L; Itsn; ITSN1; MGC134948; MGC134949; SH3 domain-containing protein 1A; SH3D1A; SH3P17; Src homology 3 domain-containing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6453 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15811 | |
| ITSN1 | |
| Recombinant protein corresponding to Human ITSN1. Recombinant protein control fragment (Product #RP-91801). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction