missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITSN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15566902
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15566902 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15566902 Supplier Invitrogen™ Supplier No. PA545703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: TFTEGEEILV TQKDGEWWTG SIGDRSGIFP SNYVKPKDQE SFGSASKSGA Sequence homology: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 93%; Rat: 79%.

This gene encodes a cytoplasmic protein which contains SH3 domains. This protein is a member of a family of proteins involved in clathrin-mediated endocytosis. Intersectin 2 is thought to regulate the formation of clathrin-coated vesicles and also may function in the induction of T cell antigen receptor (TCR) endocytosis. Alternatively spliced transcript variants have been found for this gene that encode three distinct isoforms. Additional variants have been found but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITSN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ITSN2
Gene Accession No. Q9NZM3
Gene Alias AI327390; EH and SH3 domains protein 2; EH domain and SH3 domain regulator of endocytosis 2; Eh domain, SH3 domain regulator of endocytosis 2; Ese2; intersectin 2; intersectin-2; Intersectin-2-like protein; ITSN2; KIAA1256; LOW QUALITY PROTEIN: intersectin-2; mKIAA1256; PRO2015; SH3 domain protein 1B; SH3 domain-containing protein 1B; SH3D1B; Sh3p18; SH3P18-like WASP-associated protein; SWA; SWAP
Gene Symbols ITSN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ITSN2 (aa 1001-1050).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50618
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.