missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITX3, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
ITX3 is a specific and nontoxic inhibitor of the TrioN (N-terminal GEF domain of the multidomain Trio protein) with IC50 of 76 uM
Specifications
Specifications
| Chemical Name or Material | ITX3 |
| CAS | 347323-96-0 |
| Color | Light Green |
| Purity Grade Notes | Research |
| Molecular Formula | C22H17N3OS |
| Quantity | 10 mg |
| Solubility Information | DMSO : 2 mg/mL (5.38 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(/C1=C\C2=C(C)N(C(C)=C2)C3=CC=CC=C3)N4C(S1)=NC5=CC=CC=C45 |
| Molecular Weight (g/mol) | 371.45 |
| Formula Weight | 371.45 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction