missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine hydrochloride, MedChemExpress

Product Code. 16461386
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16461386

Brand: Medchem Express HYB0162A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker that inhibits the cardiac pacemaker current (If). Ivabradine hydrochloride reduces dose-dependently heart rate without modification of blood pressure. Ivabradine hydrochloride shows anticonvulsant, anti-ischaemic and anti-anginal activity.

Ivabradine hydrochloride is a potent and orally active HCN (hyperpolarization-activated cyclic nucleotide-gated) channel blocker that inhibits the cardiac pacemaker current (If). Ivabradine hydrochloride reduces dose-dependently heart rate without modification of blood pressure. Ivabradine hydrochloride shows anticonvulsant, anti-ischaemic and anti-anginal activity.

Specifications

Chemical Name or Material Ivabradine hydrochloride
CAS 148849-67-6
Color White
Purity Grade Notes Research
Molecular Formula C27H37ClN2O5
Quantity 200 mg
Solubility Information H2O : 50 mg/mL (99.00 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (49.50 mM; Need ultrasonic)
SMILES O=C1N(CCCN(C[C@@H]2C3=CC(OC)=C(OC)C=C3C2)C)CCC4=CC(OC)=C(OC)C=C4C1.Cl
Molecular Weight (g/mol) 505.05
Formula Weight 505.05
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.