missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IVD Antibody (04), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-06536F
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
IVD Monoclonal antibody specifically detects IVD in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifica
| IVD | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 04 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IVD (AAH17202.1; His30-His423). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3712 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto