Learn More
IVD Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99159MFV500
This item is not returnable.
View return policy
Description
IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for Immunohistochemistry (Paraffin)