Learn More
IVD Mouse anti-Human, CoraLite 594, Clone: 2D12A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66032-100UL
Description
The IVD antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human IVD. This antibody recognizes human antigen. The IVD antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| IVD | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 3712 | |
| IgG2a |
| Immunofluorescence | |
| 2D12A5 | |
| Mouse | |
| Primary | |
| Human |