missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IWP-3
Description
- Inhibitor of WNT Production-3 (IWP-3) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.) IWP-3 impairs WNT pathway activity in vitro with an IC50 value of 40 nM (Chen et al.)
- Promotes cardiomyocyte differentiation from human embryonic stem cells that have been induced to mesoderm by addition of BMP4 and Activin A (Willems et al.)
Specifications
Specifications
| CAS | 687561-60-0 |
| Synonym | Inhibitor of Wnt Production-3 |
| Quantity | 10 mg |
| Molecular Formula | C22H17FN4O2S3 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?