missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ixekizumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559323
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ixekizumab is a humanized immunoglobulin G subclass 4 (IgG4) monoclonal antibody (mAb) against interleukin-17A (IL-17A) and prevents it from interacting with the IL-17A receptor. As IL-17A is a pro-inflammatory cytokine involved in inflammation and immune responses, blocking its effect is beneficial for use in inflammatory conditions. In particular, IL-17A has been found to be implicated in a variety of autoimmune diseases including Rheumatoid Arthritis and plaque psoriasis. Ixekizumab is produced by recombinant DNA technology in a recombinant mammalian cell line and purified using standard technology for bioprocessing. Ixekizumab is comprised of two identical light chain polypeptides of 219 amino acids each and two identical heavy chain polypeptides of 445 amino acids each, and has a molecular weight of 146, 158 Daltons for the protein backbone of the molecule. It is indicated for the treatment of adults with moderate-to-severe plaque psoriasis who are candidates for systemic therapy or phototherapy.
Specifications
| Ixekizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LY2439821; Taltz | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction