missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Izalontamab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554917
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA Isotype of this product is: IgG1, kappa-[scFv-heavy-lambda]2.
Izalontamab is a monoclonal antibody that targets and binds to specific antigens on cancer cells, leading to immune-mediated destruction of these cells.
Specifications
| Izalontamab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| EGFR; ERBB; ERBB1; HER1ERBB3; HER3 | |
| Recombinant Human EGFR/ERBB1/HER1 and ERBB3/HER3. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction