Learn More
GPE Scientific™ J Young O-Ring Tap Needle Valve
Product Code. 15894824
Change view
Click to view available options
Plug Bore:
1 mm
2 mm
3 mm
4 mm
6 mm
9 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore unitSize
15894824 2 mm Each
15844824 1 mm Each
15844834 3 mm Each
15854834 4 mm Each
15874834 6 mm Each
15814844 9 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15894824 Supplier GPE Scientific™ Supplier No. POR/2/NV
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young O-Ring Tap Needle Valve incorporates a PTFE piston sealed as the O-ring with a PTFE needle shut-off point.
  • The needle valve incorporates a PTFE piston sealed as the O-ring Tap with a PTFE needle shut-off point.
  • Calibrations on the barrel give an indication of flow to enable you to repeat processes.
  • Includes a single side arm
  • Available with different side arm sizes
Specifications
Type Needle valve tap with side arm
Plug Bore 2 mm
Material PTFE
Diameter (Metric) Side Arm ID: 2 mm capillary, OD: 6 mm
Includes PTFE piston