Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| For Use With (Equipment) | Vacuum manifolds and Schlenk lines |
| Includes | Fine thread tap |
| Product Type | Sample Flask |