missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAB1 Antibody (OTI2B12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01813
This item is not returnable.
View return policy
Description
JAB1 Monoclonal specifically detects JAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| JAB1 | |
| Monoclonal | |
| 0.76 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5 | |
| Mouse | |
| 37.4 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 10987 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2B12 | |
| Unconjugated | |
| Q92905 | |
| COPS5 | |
| Full length human recombinant protein of human COPS5(NP_006828) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction