missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Jagged2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111001
This item is not returnable.
View return policy
Description
Immunogen sequence: LVVIPFQFAW PRSFTLIVEA WDWDNDTTPN EELLIERVSH AGMINPEDRW KSLHFSGHVA HLELQIRVRC DENYYS Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 54%.
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
Specifications
| Jagged2 | |
| Polyclonal | |
| Unconjugated | |
| Jag2 | |
| D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y219 | |
| Jag2 | |
| Recombinant Protein corresonding to Human Jagged2. Recombinant protein control fragment (Product #RP-93665). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction