missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAGN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598615
This item is not returnable.
View return policy
Description
JAGN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Endoplasmic reticulum transmembrane protein involved in vesicle-mediated transport, which is required for neutrophil function. Required for vesicle-mediated transport; it is however unclear whether it is involved in early secretory pathway or intracellular protein transport. Acts as a regulator of neutrophil function, probably via its role in vesicle-mediated transport: required for defense against fungal pathogens and for granulocyte colony-stimulating factor (GM-CSF) signaling pathway; possibly by regulating glycosylation and/or targeting of proteins contributing to the viability and migration of neutrophils.
Specifications
| JAGN1 | |
| Polyclonal | |
| Unconjugated | |
| JAGN1 | |
| 5830427H10Rik; GL009; JAGN1; jagunal homolog 1; jagunal homolog 1 (Drosophila); protein jagunal homolog 1; SCN6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84522 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N5M9 | |
| JAGN1 | |
| Recombinant Human Protein jagunal homolog 1 protein (1-39AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction