missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jak2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22975F
This item is not returnable.
View return policy
Description
Jak2 Polyclonal antibody specifically detects Jak2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Jak2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2 | |
| The immunogen recognized by this antibody maps to a region between residue 1082 and 1132 of human Janus kinase 2 using the numbering given in entry NP_004963.1 (GeneID 3717). | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| 3717 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction