Learn More
Jak2, p Tyr1007 Antibody (SR1129), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21579-100ul
 View more versions of this product
Product Code. 18651864
  • € 449.00 / 100µL
Description
Jak2 Monoclonal antibody specifically detects Jak2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Serum
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Jak2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
SR1129
Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:50, Serum 1:50-1:200
EC 2.7.10, EC 2.7.10.2, JAK-2, Janus kinase 2Janus kinase 2 (a protein tyrosine kinase), JTK10, tyrosine-protein kinase JAK2
A synthesized peptide derived from human phospho-Jak2 [Tyr1007, Tyr1008] (Uniprot #: O60674)
100 μg
Cell Cycle and Replication, Phospho Specific, Signal Transduction
3717
Store at -20 degrees C. Avoid freeze/thaw cycles.