missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAKMIP2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41202DL594
This item is not returnable.
View return policy
Description
JAKMIP2 Polyclonal antibody specifically detects JAKMIP2 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| JAKMIP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| DyLight 594 | |
| CTCL tumor antigen HD-CL-04, JAMIP2KIAA0555Jak and microtubule interacting protein 2, janus kinase and microtubule interacting protein 2, janus kinase and microtubule-interacting protein 2, NECC1, neuroendocrine long coiled-coil 1, Neuroendocrine long coiled-coil protein 1 | |
| Antibody was raised against a 16 amino acid peptide near the center of human JAKMIP2. The immunogen is located within amino acids 250 - 300 of JAKMIP2. Amino Acid Squence: RAGDGSEHCSSPDLR | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 9832 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction