missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-A Antibody (H202-106), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65340FR
This item is not returnable.
View return policy
Description
JAM-A Monoclonal specifically detects JAM-A in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| JAM-A | |
| Monoclonal | |
| DyLight 680 | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 50848 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| H202-106 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| F11R | |
| MTE1/MTE2 stromal cell lines. | |
| 0.25 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction