missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-A Antibody (OTI3H3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71876AF647
This item is not returnable.
View return policy
Description
JAM-A Monoclonal specifically detects JAM-A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| JAM-A | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| F11R | |
| Full length recombinant protein of human JAM-A (NP_653087) produced in HEK293T cells. | |
| 0.1 mL | |
| 50848.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3H3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CD321 antigen, F11 receptor, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1KAT, Platelet adhesion molecule 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction