missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-B/VE-JAM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | JAM-B/VE-JAM |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680659
|
Novus Biologicals
NBP2-68604-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604068
|
Novus Biologicals
NBP2-68604 |
100 μg |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JAM-B/VE-JAM Polyclonal antibody specifically detects JAM-B/VE-JAM in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| JAM-B/VE-JAM | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS (pH 7.2) and 40% Glycerol | |
| 58494 | |
| IgG | |
| Protein A purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GVCYAQRKGYFSKETSFQKSNSSSKATTMS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title