missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-C Antibody (208212), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11891MFV610
This item is not returnable.
View return policy
Description
JAM-C Monoclonal antibody specifically detects JAM-C in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| JAM-C | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Signal Transduction | |
| 83700 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 208212 | |
| Flow Cytometry, CyTOF-ready | |
| JAM-2, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C | |
| Mouse myeloma cell line NS0-derived recombinant human JAM-C, Val32-Asn241(Ala149Pro), Accession # Q9BX67 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction