missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JAML Polyclonal Antibody
GREENER_CHOICE

Product Code. 17122124 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17122124 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17122124 Supplier Invitrogen™ Supplier No. PA5114263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VCATILLLPV LILIVKKTCG NKSSVNSTVL VKNTKKTNPE MKEKPCHFER For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JAML
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene JAML
Gene Accession No. Q86YT9
Gene Alias adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387
Gene Symbols JAML
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of human JAML.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.