Learn More
Tocris Bioscience™ Janelia Fluor™ 552, Haloalkane
Description
Application: live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, light sheet microscropy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). In vivo functional imaging of Ca2+ responses (flies/mice); multiplexed functional imaging of neurons and astrocytes in zebrafish larvae; quantification of Ca2+ concentration using fluorescence lifetime imaging microscopy (FLIM).
Properties and Photophysical Data: Stable to fixation. Cell permeable. KD = 87 ? 5. In the presence of EGTA (Ca2+ chelator) excitation and emission maxima (λ) are 561 nm and 559-578 nm, respectively; quantum yield = 0.05; extinction coefficient = 73,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); brigthness = 3.6 mM-1cm-1. In the presence of saturating Ca2+, excitation and emission maxima (λ) are 562 nm and 565-582 nm, respectively; quantum yield = 0.25; extinction coefficient = 78,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); brigthness = 19.5 mM-1cm-1.
Janelia Fluor® is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-6-(azetidin-1-yl)-2,7-difluoro-3H-xanthen-9-yl)-4-((2-(2-((6-chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)benzoate |
| CAS | 2634129-34-1 |
| Quantity | 10 μg |
| Target | Self-labeling Tags |
| Molecular Formula | C37H40ClF2N3O6 |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.