missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Janelia Fluor™ 635, Haloalkane

Product Code. 30241044
Click to view available options
:
10 μg
This item is not returnable. View return policy

Product Code. 30241044

Brand: Tocris Bioscience™ 8808/10U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell-permeable fluorogenic fluorescent dye with a chloroalkane handle. Application: live-cell imaging as a self-labeling tag substrate

Key Information: Janelia Fluor™ 635, Haloalkane is a red cell-permeable fluorogenic fluorescent dye with a chloroalkane handle.

Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, light sheet microscropy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). Stable to fixation. Cell permeable.

Properties and Photophysical Data: Janelia Fluor™ 635, Haloalkane shows a high degree of fluorogenicity. Excitation and emission maxima (λ) are 635 nm and 652 nm, respectively; quantum yield = 0.56; extinction coefficient = 167,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); lactone-zwitterion equilibrium constant (KL-Z) <0.0001. Turn-on ratio in vitro A/A0 = 113, F/F0 = 67. Exhibits low non-specific background staining.

Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-((2-(2-((6-Chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)-2-(3-(3-fluoroazetidin-1-ium-1-ylidene)-7-(3-fluoroazetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)benzoate
CAS 1811539-66-8
Quantity 10 μg
Target Janelia Fluor Dyes
Molecular Formula C39H46ClF2N3O5Si
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.