missing translation for 'onlineSavingsMsg'
Learn More

JANEX-1, MedChemExpress

Product Code. 16409157
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409157

Brand: Medchem Express HY155085mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2.

JANEX-1 (WHI-P131) is a potent and specific JAK3 inhibitor (estimated Ki=2.3 μM). JANEX-1 (WHI-P131) shows potent JAK3-inhibitory activity (IC50 of 78 μM), does not inhibit JAK1 and JAK2.

Specifications

Chemical Name or Material JANEX-1
CAS 202475-60-3
Color Gray
Purity Grade Notes Research
Molecular Formula C16H15N3O3
Quantity 5 mg
Synonym WHI-P131 Jak3 inhibitor I
Solubility Information DMSO : ≥ 100 mg/mL (336.35 mM)
SMILES OC1=CC=C(NC2=C3C=C(OC)C(OC)=CC3=NC=N2)C=C1
Molecular Weight (g/mol) 297.31
Formula Weight 297.31
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.