missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Japanese Encephalitis Virus NS1 Monoclonal Antibody (GT1410)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA535916
This item is not returnable.
View return policy
Description
Japanese Encephalitis Virus NS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Japanese encephalitis virus (JEV) is a flavivirus related to dengue, yellow fever and West Nile viruses, and is spread by mosquitoes. JEV is the main cause of viral encephalitis in many countries of Asia with an estimated 68000 clinical cases every year.
Specifications
| Japanese Encephalitis Virus NS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 20% glycerol and no preservative; pH 7 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT1410 | |
| Unconjugated | |
| Japanese Encephalitis | |
| Recombinant protein encompassing a sequence within the center region of NS1 (JEV) (Japanese Encephalitis Virus strain Jaoars982). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction