missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JARID1A, Mouse anti-Human, Clone: 9A6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB6751.100ug
This item is not returnable.
View return policy
Specifications
| JARID1A | |
| Monoclonal | |
| Unconjugated | |
| In PBS (50% glycerol) | |
| KDM5A/RBBP2/RBP2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is specific to human and mouse KDM5A. It can detect endogenous levels of KDM5A. | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot | |
| 9A6 | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| jumonji, AT rich interactive domain 1A | |
| JARID1A | |
| A synthetic peptide corresponding to amino acids 1416-1434 of human KDM5A. | |
| RUO | |
| 5927 | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction