missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JARID2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16396734
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16396734 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16396734 Supplier Invitrogen™ Supplier No. PA578032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: human JARID2-transfected 293T, fetal mouse forebrain Predicted reactivity: Mouse (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Jumonji (JARID2) is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. Also, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JARID2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene JARID2
Gene Accession No. Q62315, Q92833
Gene Alias JARD2 antibody; Jarid2; Jmj; jumonji; jumonji and AT-rich interaction domain containing 2; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji, AT rich interactive domain 2 protein; jumonji/ARID domain-containing protein 2; jumonji-like protein; LOW QUALITY PROTEIN: protein Jumonji; protein Jumonji
Gene Symbols JARID2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human JARID2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16468, 3720
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.