missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JARID2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585539

Product Code. 16342475

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (82%). Positive Control: human JARID2-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Jumonji (JARID2) is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. Also, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JARID2
Polyclonal
Unconjugated
JARID2
JARD2 antibody; Jarid2; Jmj; jumonji; jumonji and AT-rich interaction domain containing 2; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji, AT rich interactive domain 2 protein; jumonji/ARID domain-containing protein 2; jumonji-like protein; LOW QUALITY PROTEIN: protein Jumonji; protein Jumonji
Rabbit
Antigen affinity chromatography
RUO
3720
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q92833
JARID2
Recombinant protein encompassing a sequence within the N-terminus region of human JARID2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.