Learn More
JIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14568-1-AP-150UL
 View more versions of this product
Product Code. 16873264
  • € 410.00 / 150µL
Description
The protein encoded by this gene is a regulator of the pancreatic beta-cell function. It is highly similar to JIP-1, a mouse protein known to be a regulator of c-Jun amino-terminal kinase (Mapk8). This protein has been shown to prevent MAPK8 mediated activation of transcription factors, and decrease IL-1 beta and MAP kinase kinase 1 (MEKK1) induced apoptosis in pancreatic beta cells. This protein also functions as a DNA-binding transactivator of the glucose transporter GLUT2. RE1-silencing transcription factor (REST) is reported to repress the expression of this gene in insulin-secreting beta cells. This gene is found to be mutated in a type 2 diabetes family, and thus is thought to be a susceptibility gene for type 2 diabetes.
Specifications
Specifications
JIP1
Polyclonal
Unconjugated
MAPK8IP1
IB 1, IB1, Islet brain 1, JIP 1, JIP1, JNK interacting protein 1, MAPK8IP1, PRKM8IP
Rabbit
Antigen Affinity Chromatography
RUO
116457, 19099, 9479
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9R237, Q9UQF2, Q9WVI9
Mapk8ip1
JIP1 Fusion Protein Ag6150
150 μL
Primary
Human, Mouse, Rat