missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101157
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total JIP2.
The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.
Specifications
| JIP2 | |
| Polyclonal | |
| Unconjugated | |
| MAPK8IP2 | |
| 3230402N03Rik; AI847694; AW049958; C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB 2; Ib2; IB-2; Islet brain 2; islet-brain 2; islet-brain-2; JIP 2; JIP2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; Mapk8ip2; mitogen activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8-interacting protein 2; PRKM8IPL; R75148; si:dkey-180h12.2; si:dkey-181m21.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23542, 315220, 561262, 60597 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| G3V9M2, Q13387, Q9ERE9 | |
| MAPK8IP2 | |
| A synthesized peptide derived from human MAPK8IP2(Accession Q13387), corresponding to amino acid residues V581-V631. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction