missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD1B Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77071AF750
This item is not returnable.
View return policy
Description
JMJD1B Polyclonal antibody specifically detects JMJD1B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| JMJD1B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| C5orf7, EC 1.14.11, EC 1.14.11.-, JHDM2B, JmjC domain-containing histone demethylation protein 2B, JMJD1B, jumonji domain containing 1B, Jumonji domain-containing protein 1B, KIAA1082chromosome 5 open reading frame 7, lysine (K)-specific demethylase 3B, lysine-specific demethylase 3B, NET22,5qNCA, Nuclear protein 5qNCA | |
| Antibody was raised against a 20 amino acid synthetic peptide from near the center of human JMJD1B. The immunogen is located within amino acids 1050 - 1100 of JMJD1B. Amino Acid Squence: YRLRKSRPRSETEEMGDEE | |
| 0.1 mL | |
| Cancer | |
| 51780 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction