missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD1C Antibody (3A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00221037-M02
This item is not returnable.
View return policy
Description
JMJD1C Monoclonal antibody specifically detects JMJD1C in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| JMJD1C | |
| Monoclonal | |
| Unconjugated | |
| NP_004232 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 221037 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3A8 | |
| In 1x PBS, pH 7.4 | |
| DKFZp761F0118, EC 1.14.11, EC 1.14.11.-, FLJ14374, JHDM2C, jumonji domain containing 1C, Jumonji domain-containing protein 1C, KIAA1380TR-interacting protein 8, probable JmjC domain-containing histone demethylation protein 2C, RP11-10C13.2, thyroid hormone receptor interactor 8, thyroid receptor interacting protein 8, Thyroid receptor-interacting protein 8, TRIP8, TRIP-8 | |
| JMJD1C (NP_004232, 2 a.a. ∽ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IVMNDQVLEPQNVDPSMVQMTFLDDVVHSLLKGENIGITSRRRSRANQNVNAVHSHYTRAQANSPRPAMNSQAAVPKQNTHQQQQQRSIRPNKRKGSD | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction