missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585519
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Positive Control: HeLa nuclear extract, Rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Specifications
| JMJD2A | |
| Polyclonal | |
| Unconjugated | |
| KDM4A | |
| D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 313539, 9682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.84 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75164 | |
| KDM4A | |
| Recombinant protein encompassing a sequence within the C-terminus region of human KDM4A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction