Learn More
Invitrogen™ JMJD2A Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115459
Product Code. 17218243
  • € 506.00 / 100µL
Description
Antibody detects endogenous levels of total KDM4A.
JMJD2A is a member of the Jumonji domain 2 (JMJD2) family and a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
JMJD2A
Polyclonal
Unconjugated
KDM4A
D4Ertd222e; JHDM3A; jmjC domain-containing histone demethylation protein 3A; JMJD2; JMJD2A; jumonji C domain-containing histone demethylase 3A; jumonji domain containing 2; jumonji domain containing 2A; jumonji domain-containing protein 2A; KDM4A; KIAA0677; lysine (K)-specific demethylase 4A; lysine demethylase 4A; lysine-specific demethylase 4A; mKIAA0677; TDRD14A; tudor domain containing 14A
Rabbit
Affinity chromatography
RUO
230674, 313539, 9682
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
O75164, Q8BW72
KDM4A
A synthesized peptide derived from human KDM4A(Accession O75164), corresponding to amino acid residues R574-L624.
100 μL
Primary
Human, Mouse, Rat