missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD2B Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74604JF646
This item is not returnable.
View return policy
Description
JMJD2B Polyclonal antibody specifically detects JMJD2B in Human, Chicken samples. It is validated for Western Blot
Specifications
| JMJD2B | |
| Polyclonal | |
| Western Blot | |
| EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B | |
| The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human jumonji domain containing 2B using the numbering given in entry NP_055830.1 (GeneID 23030). | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 23030 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction