missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD2B Recombinant Rabbit Monoclonal Antibody (JE36-58)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550831
This item is not returnable.
View return policy
Description
Positive control: HeLa cell lysate, HCT116 cell lysate, SW480 cell lysate, mouse testis tissue lysate, rat testis tissue lysates, human breast carcinoma tissue, HeLa, SW480. Predicted band size: 122 kDa Subcellular Location: Nucleus.
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt].
Specifications
| JMJD2B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O94953, Q91VY5 | |
| KDM4B | |
| Synthetic peptide within Human KDM4B aa 97-146/1096. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE36-58 | |
| Unconjugated | |
| KDM4B | |
| 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 193796, 23030 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction