Learn More
Invitrogen™ JMJD2C Monoclonal Antibody (OTI5B9)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA524956
Product Code. 15914564
  • € 574.00 / 100µL
Description
JMJD2C Monoclonal Antibody for Western Blot, IHC (P), ICC/IF
Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 'Lys-9' and H3 'Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
JMJD2C
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H3R0
KDM4C
Full-length protein expressed in 293T cell transfected with human KDM4C expression vector.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI5B9
Unconjugated
KDM4C
2410141F18Rik; AA517467; bA146B14.1; GASC1; GASC-1 protein; gene amplified in squamous cell carcinoma 1; gene amplified in squamous cell carcinoma 1 protein; JHDM3C; JmjC domain-containing histone demethylation protein 3C; JMJD2C; jumonji domain containing 2C; jumonji domain-containing protein 2C; Kdm4c; KIAA0780; lysine (K)-specific demethylase 4C; lysine demethylase 4C; lysine-specific demethylase 4C; RP11-169L18.1; TDRD14C; tudor domain containing 14C
Mouse
Affinity Chromatography
RUO
23081
-20°C, Avoid Freeze/Thaw Cycles