missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD4 Antibody (JMJD4/32b/H12), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50425AF594
This item is not returnable.
View return policy
Description
JMJD4 Monoclonal specifically detects JMJD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.
Specifications
| JMJD4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| JMJD4 | |
| Recombinant protein | |
| 0.1 mL | |
| Primary | |
| Human |
| Western Blot, ELISA, Immunohistochemistry, KnockDown | |
| JMJD4/32b/H12 | |
| Western Blot, ELISA, Immunohistochemistry | |
| FLJ12517, jumonji domain containing 4, jumonji domain-containing protein 4 | |
| Mouse | |
| Protein A purified | |
| Epigenetics | |
| 65094 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction