missing translation for 'onlineSavingsMsg'
Learn More

JNJ-40411813, MedChemExpress

Product Code. p-200100981
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452867

Brand: Medchem Express HY15748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-40411813 (ADX-71149) is a novel positive allosteric modulator of the metabotropic Glutamate 2 receptor (mGlu2R) with EC50 of 147 nM.

JNJ-40411813 (ADX-71149) is a novel positive allosteric modulator of the metabotropic Glutamate 2 receptor (mGlu2R) with EC50 of 147 nM.

Specifications

Chemical Name or Material JNJ-40411813
CAS 1127498-03-6
Color White
Purity Grade Notes Research
Molecular Formula C20H25ClN2O
Quantity 50 mg
Synonym ADX-71149
Solubility Information DMSO : 25 mg/mL (72.49 mM; Need ultrasonic)
SMILES O=C1C(Cl)=C(N2CCC(C3=CC=CC=C3)CC2)C=CN1CCCC
Molecular Weight (g/mol) 344.88
Formula Weight 344.88
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.