missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1 Antibody (OTI4E3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71256AF700
This item is not returnable.
View return policy
Description
JNK1 Monoclonal specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| JNK1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAPK8 | |
| Recombinant protein fragment corresponding to amino acids 1-384 of human JNK1 (NP_002741) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4E3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Hypoxia, Immunology, Innate Immunity, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5599.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction