missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JNK1 Monoclonal Antibody (4A2G12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 359800
This item is not returnable.
View return policy
Description
JNK1 Monoclonal Antibody for Western Blot, ICC/IF
MAPK8 (JNK1) is activated by the presence of misfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity. It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
Specifications
| JNK1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P45983 | |
| MAPK8 | |
| Recombinant GST-fusion protein of JNK1. | |
| 100 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 4A2G12 | |
| Unconjugated | |
| MAPK8 | |
| AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; zgc:112379 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5599 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction