Learn More
Eurofins DiscoverX JNK3/SAPK1b Protein, Active
Description
c-Jun N-terminal kinases (JNKs), a stress-activated protein kinase (SAPK), is involved in cellular responses to environmental stresses. JNK kinases can be activated by stimuli such as UV light, radiation, protein synthesis inhibitors, ceramide, DNA-damaging drugs, chemopreventive drugs, TNF-a, and interleukin 1. JNK binds to an NH2-terminal region of ATF2 and c-Jun and phosphorylates two sites within the activation domain of each transcription factor. Three JNK genes (Jnk1, -2 and -3) have been identified in humans; however, splice variants result in a total of 10 isoforms. Inhibition of JNK3 (MAPK10, Mitogen-activated protein kinase 10, Stress-activated protein kinase JNK3) may provide clinical benefit in diseases as diverse as immune inflammatory (joint, bowel, pulmonary), ischemic injury (brain, cardiac) and neurodegenerative (Alzheimer's, Parkinson's) diseases.
The JNK3 protein sequence is identical to the human JNK3/SAPK1b described inGenBank NM_138980. The DNA sequence, however, is more closely related to the rat cDNA described inGenBank NM_012806.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 53 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥77% |
| Protein | JNK3/SAPK1b, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.