missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Josephin1 Antibody (OTI3B11), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71506AF594
This item is not returnable.
View return policy
Description
Josephin1 Monoclonal antibody specifically detects Josephin1 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Josephin1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 3.4.19.12, Josephin domain containing 1, Josephin domain-containing 1, Josephin-1, JSPH1, KIAA0063dJ508I15.2 | |
| Full length recombinant protein of human Josephin1 (NP_055691) produced in HEK293T cells. | |
| 0.1 mL | |
| Cell Biology | |
| 9929 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction