missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Josephin1 Antibody (OTI3B11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71506CL1
This item is not returnable.
View return policy
Description
Josephin1 Monoclonal antibody specifically detects Josephin1 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Josephin1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.4.19.12, Josephin domain containing 1, Josephin domain-containing 1, Josephin-1, JSPH1, KIAA0063dJ508I15.2 | |
| Full length human recombinant protein of human JOSD1 (NP_055691) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology | |
| 9929 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI3B11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction