missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JPH1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15720621 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15720621 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15720621 Supplier Invitrogen™ Supplier No. PA552668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PQSKYSGRHH IPNPSNGELH SQYHGYYVKL NAPQHPPVDV EDGDGSSQSS SALVHKPSAN KWSPSKSVTK PVAKESKAEP KAKKSELAIP KNPASNDSCP AL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

Junctional complexes between the plasma membrane and endoplasmic/sarcoplasmic reticulum are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. The protein encoded by this gene is a component of junctional complexes and is composed of a C-terminal hydrophobic segment spanning the endoplasmic/sarcoplasmic reticulum membrane and a remaining cytoplasmic domain that shows specific affinity for the plasma membrane. This gene is a member of the junctophilin gene family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Jph1
Gene Accession No. Q9HDC5
Gene Alias CMT2K; ENSMUSG00000054314; JP1; JP-1; Jph1; junctophilin 1; junctophilin type 1; junctophilin type1; Junctophilin-1; mitsugumin72
Gene Symbols Jph1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human JPH1. Recombinant protein control fragment (Product #RP-90044).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.