missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JPH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25979-1-AP-150UL
This item is not returnable.
View return policy
Description
Junctional complexes between the plasma membrane and endoplasmic/sarcoplasmic reticulum are a common feature of all excitable cell types and mediate cross talk between cell surface and intracellular ion channels. The protein encoded by this gene is a component of junctional complexes and is composed of a C-terminal hydrophobic segment spanning the endoplasmic/sarcoplasmic reticulum membrane and a remaining cytoplasmic domain that shows specific affinity for the plasma membrane. This gene is a member of the junctophilin gene family.Specifications
| JPH1 | |
| Polyclonal | |
| Unconjugated | |
| JPH1 | |
| JP 1, JP1, JPH1, junctophilin 1, Junctophilin type 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56704, 57339 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ET80, Q9HDC5 | |
| JPH1 | |
| JPH1 Fusion Protein Ag23270 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction