missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JPH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41095
This item is not returnable.
View return policy
Description
JPH4 Polyclonal specifically detects JPH4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| JPH4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| JPH4 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human JPH4. The immunogen is located within amino acids 410 - 460 of JPH4. Amino Acid Squence: LEAPFRRPRQDSEGSDT | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA | |
| JP4, JP-4, JPHL1, Junctophilin 4, Junctophilin Like 1, junctophilin-4, Junctophilin-Like 1 Protein, KIAA1831 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84502 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction