Learn More
Tocris Bioscience™ JPS036
Description
JPS036 is a selective HDAC3 Degrader (PROTAC™) (DC50 = 440 nM; Dmax = 77 %). JPS036 comprises a benzamide-based compound joined by a linker to a Von Hippel Lindau (VHL) E3 ligase ligand.
Negative control JPS016NC also available.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-N-[2-[[12-[[4-[(2-Aminophenyl)carbamoyl]phenyl]amino]-12-oxododecyl]oxy]-4-(4-methylthiazol-5-yl)benzyl]-1-[(S)-2-(1-fluorocyclopropan-1-ylcarbonylamino)-3,3-dimethylbutanoyl]-4-hydroxypyrrolidine-2-carboxamide |
| CAS | 2669785-85-5 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C51H66FN7O7S |
| Purity | 0.98 |